Imprimer
Safety burners with stopcock
Fournisseur: USBECK Laborgeräte
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1340 1341
451-1369EA 168 EUR
451-1369 USBE1341
Safety burners with stopcock
Appareils chauffants Brûleurs à gaz
This safety burner provides protection against uncontrolled escape of gas in laboratories. If the flame is extinguished, the gas supply is interrupted and no unburnt gas can escape.
  • Safety pilot flame
  • 90° stopcock
  • With air regulation and thermocouple