Imprimer
Essuyeurs à usage général, Multitex® Ultra z 60
Fournisseur: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10142-60BOX 10142-60W 11142-60B 11142-60W
115-2472EA 335 EUR
115-2472 ZVGA10142-60W ZVGA11142-60B ZVGA11142-60W
Essuyeurs à usage général, Multitex® Ultra z 60
Lingettes Lingettes et essuie-mains à usage unique Lingettes en papier
These high-quality non woven wipes are suitable for moist and dry cleaning applications.
  • Disposable
  • Very soft and durable
  • Can absorb large amounts of liquid
  • Tear-resistant, solvent-resistant and food-safe
Ideal for use in sensitive work areas, such as precision mechanics, electrical and automotive industries, and in paint shops.