Imprimer
Chaussures de travail, à lacets, Uni6, 1760
Fournisseur: Abeba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1760/35 1760/47 1760/48 1760/36 1760/40 1760/41 1760/37 1760/38 1760/39 1760/42 1760/43 1760/44 1760/45 1760/46
ABEB1760/35EA 115 EUR
ABEB1760/35 ABEB1760/47 ABEB1760/48 ABEB1760/36 ABEB1760/40 ABEB1760/41 ABEB1760/37 ABEB1760/38 ABEB1760/39 ABEB1760/42 ABEB1760/43 ABEB1760/44 ABEB1760/45 ABEB1760/46
Chaussures de travail, à lacets, Uni6, 1760
Chaussures et bottes
Anti-static shoe made from functional leather with honey comb pattern, non-slip sole and fuel resistant foamed PU sole.
  • Water-repellent upper
  • Breathable lining with Silver Point
  • Replaceable Soft Comfort insole M
  • Foamed PU midsole
  • Sanitized® treatment reduces odour and bacterial growth
  • Protective steel toecap
Colour: White
Certifications: DGUV 112-191; EN 20347:2012; S1, SRC.