Imprimer
VWR®, Filter Tips for Rainin® LTS® Pipettes, Low Retention
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
732-3339EA 156 EUR
732-3339 732-3340 732-3343
VWR®, Filter Tips for Rainin® LTS® Pipettes, Low Retention
Pipette Tips
These low retention filter tips are made with a special resin that reduces the potential for sample retention inside the tip. They help users reduce sample loss when performing protein research. These tips fit Rainin® LTS® pipettes.
  • Sustainable design based on virgin resins
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins, human DNA and trace metals
  • Lot certified to be free of protease and nuclease
  • Tested for ATP
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.
Packaging options:
Racks (96 tips per rack): These colour coded racks, with smooth rounded edges, have heavy hook hinges that are extremely durable and an ergonomic snap latch that won’t catch gloves. Strong enough to be autoclaved many times. The separate rack insert includes a bevelled top and extra sturdy surface for multi-channel pipetting.
Next Generation refills: Next Generation refill systems refill VWR and other brands of pipette tip racks. Their low carbon footprint makes them an ideal solution for waste reduction goals in the lab. Users don’t need gloves to refill tip racks - Next Generation refills racks without users having to touch the new tips.