Imprimer
Cannula filters
Fournisseur: QUALITY LAB ACCESSORIES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FIL020-HR-1000 FIL010-HR
QLAAFIL020-HR-1000EA 2780 EUR
QLAAFIL020-HR-1000 QLAAFIL010-HR
Cannula filters
Filtres Tubes filtrants
Cannula filters for dissolution sampling methods.
  • Filters made from inert HDPE
  • Made of ultra-high molecular weight sintered PE (UHMWPE)
  • All formulations have been verified by Porisometer testing
Specifically formulated to provide the specified micron ratings.