Imprimer
Scalpel handles, Major
Fournisseur: SWANN MORTON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
639 640
SWAN639EA 389 EUR
SWAN639 SWAN640
Scalpel handles, Major
Bistouris et pièces détachées Manches de bistouri
The heavy duty handles are made from stainless steel.
  • Available as a short and long version
  • Long handle provides additional reach and depth of surgical field
Fit the Swann Major orthopaedic blades S, T, U, V, W, X, Y and Z as well as the cervical biopsy blade.