Imprimer
Protease Inhibitor Cocktail, Animal-Free, Calbiochem®
Marques : Merck Millipore Calbiochem
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
535140-1 535140-SET
535140-1EA 108 EUR
535140-1 535140-SET
Protease Inhibitor Cocktail, Animal-Free, Calbiochem®
Suppléments et additifs pour culture cellulaire
Calbiochem® protease inhibitor cocktail set III, animal-free, is a cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine and serine proteases and aminopeptidases.
Recommended to use with mammalian cell and tissue extracts.