Imprimer
ESD safety shoes, lace-up, uvex 2, vibram®, 6528
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6528.2/51 6528.2/52 6528.2/47 6528.2/48 6528.2/39 6528.2/40 6528.2/50 6528.2/37 6528.2/38 6528.2/43 6528.2/44 6528.2/45 6528.2/46 6528.2/35 6528.2/36 6528.2/49 6528.2/41 6528.2/42 6528.4/48 6528.4/49 6528.4/50 6528.4/51 6528.4/52 6528.4/35 6528.4/36 6528.4/37 6528.4/38 6528.4/39 6528.4/40 6528.4/41 6528.4/42 6528.4/43 6528.4/44 6528.4/45 6528.4/46 6528.4/47
114-8111EA 164 EUR
114-8111 114-8112 114-8107 114-8108 114-8099 114-8100 114-8110 114-8097 114-8098 114-8103 114-8104 114-8105 114-8106 114-8095 114-8096 114-8109 114-8101 114-8102 UVEX6528.4/48 UVEX6528.4/49 UVEX6528.4/50 UVEX6528.4/51 UVEX6528.4/52 UVEX6528.4/35 UVEX6528.4/36 UVEX6528.4/37 UVEX6528.4/38 UVEX6528.4/39 UVEX6528.4/40 UVEX6528.4/41 UVEX6528.4/42 UVEX6528.4/43 UVEX6528.4/44 UVEX6528.4/45 UVEX6528.4/46 UVEX6528.4/47
ESD safety shoes, lace-up, uvex 2, vibram®, 6528
Chaussures et bottes
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water repellent high-tech microvelour upper, soft padded tongue and collar, and a dual-density slip-resistant VIBRAM® rubber sole resistant to chemicals, oils and gasoline.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction to eliminate pressure points
  • Lightweight two-layer sole with excellent shock absorption outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuffcap for additional protection
  • Flexible, metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including the chemical, transport, logistics, energy and mechanical engineering industries.
Colour: Black/orange
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, HI, HRO, SRC