Imprimer
Crucibles, cylindrical, flat base, aluminum oxide (Al-24)
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
32961.KF 32962.KF 32963.KF 32964.KF 32965.KF 32966.KF 32969.KF
32961.KFEA 74.8 EUR
32961.KF 32962.KF 32963.KF 32964.KF 32965.KF 32966.KF 32969.KF
Crucibles, cylindrical, flat base, aluminum oxide (Al-24)
Creusets
These crucibles are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
  • 1950 °C (max service temp in air)