Imprimer
Avantor® Prevail C18-Select, HPLC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICH99316EA 1020 EUR
HICH99316 HICH99300 HICH99301 HICH99309 HICH99313 HICH99302 96691/N 99119/N
Avantor® Prevail C18-Select, HPLC Columns
Colonnes pour chromatographie
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
Same benefits as Prevail™ C18, but slightly more retentive.
Informations sur les accessoires: Use guard cartridges with holder (80101/N) and column coupler (HI-081).