Imprimer
Vydac 238TP C18, HPLC columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
238TP510EA 0 EUR
238TP510 238TP53 238TP54 238GK52/N 238GK54/N
Vydac 238TP C18, HPLC columns
Colonnes pour chromatographie
Vydac TP is the first range of wide pore media phases developed in the Vydac range and it is the industry's standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
These first generation monomeric C18 phase columns are recommended for the same applications as 218TP but offers alternative C18 selectivity.
* Use with holder 80101/N and column coupler HI-081.
Vydac is a registered trademark of W.R.Grace and Co.