Imprimer
Vydac Everest C18, HPLC columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
238EV5.07515EA 1370 EUR
238EV5.07515 238EV5C0110 238EV5.1515 238EV5.1525 238EV5215 238GCC510EV 238EV1022 238GD54EV/N 238GD52EV/N 238GK54EV/N
Vydac Everest C18, HPLC columns
Colonnes pour chromatographie
Everest columns have unique selectivity and sensitivity, which are the result of bonding technology that improves C18 surface coverage and deactivates residual silanols. Previously, the 300 Å C18 chemistries had carbon coverage in the 2,8 to 3,6 µmol/m² range. Everest C18 coverage is in excess of 4 µmol/m² and approximates the theoretical limit based on surface area. The increased shielding of the base silica increases column lifetime and reduces the amount of TFA required to shield the silica.
  • Unique selectivity for hydrophilic and hydrophobic peptides
  • High resolution for complex peptide digests
  • Excellent sensitivity with little or no TFA in mobile phase
Vydac 238EV (C18) offers maximum surface coverage for the highest resolution of complex samples. It is recommended for the analysis of complex enzymatic digests (>12 proteins).
* Use with holder GCH10 and column coupler HI-081.
** Use with holder 80101/N and column coupler HI-081.