Imprimer
Handheld digital microscopes, Dino-Lite Special Light, UV
Fournisseur: Dino-Lite / IDCP B.V.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AM4113T-FV2W AM4113T-FVW AM4113T-VW AM4013MT-FVW AM4113FVT AM4113FVT2 AM4113TL-FVW AM4115T-FUW AM4115T-FVW AM4115TL-FVW AM4115-FUT AM4115-FVT AM7115MT-FUW
IDCPAM4113T-FV2WEA 493 EUR
IDCPAM4113T-FV2W IDCPAM4113T-FVW IDCPAM4113T-VW IDCPAM4013MT-FVW IDCPAM4113FVT IDCPAM4113FVT2 IDCPAM4113TL-FVW IDCPAM4115T-FUW IDCPAM4115T-FVW IDCPAM4115TL-FVW IDCPAM4115-FUT IDCPAM4115-FVT IDCPAM7115MT-FUW
Handheld digital microscopes, Dino-Lite Special Light, UV
Microscopes
These handheld microscopes feature ultraviolet (UV) light or a combination between UV and white light.
  • High speed USB 2.0 output
  • Metal housing or composite housing; metal models are ESD-safe
  • Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
  • Exchangeable caps for more flexibility (some models only)
Informations de livraison: Supplied with carry pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), user manual, Quick guide and calibration target.