Imprimer
Cut resistant gloves, JACKSON SAFETY* G60 PURPLE NITRILE*
Fournisseur: KIMBERLY CLARK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
97432 97433 97430 97431 97434
112-4860EA 297 EUR
112-4860 112-4861 112-4858 112-4859 112-1236
Cut resistant gloves, JACKSON SAFETY* G60 PURPLE NITRILE*
Gants
These ambidextrous gloves made from Ultra-high-molecular-weight polyethylene (HMPE) fibres provide protection against lacerations while enhancing grip and dexterity.
  • Purple foam nitrile coated fingertips and flexible dotted palm for enhanced grip
  • HMPE fibre enhances breathability and protection
  • Knitted fibre enhances breathability and comfort
  • Ambidextrous design improves cost-in-use
  • Washable
  • Latex-free
Typical applications include: Metal fabrication, glass handling, handling sharp parts, automotive assembly, ceramics.
Certifications: EN 388 (434X); Cat. II