Imprimer
Lingettes sèches pour salles propres, B'Wipes 6000
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
115-1821EA 90.4 EUR
115-1821 115-1823
Lingettes sèches pour salles propres, B'Wipes 6000
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
2 ply polyester sealed-border wipe for use in ISO 3 cleanrooms.
  • Quilted polyester wipe, constructed from 2 plies, with sealed borders
  • Laundered, with sealed borders, for excellent cleanliness
  • 2 ply quilted, for extreme absorption
  • Ideal for spill control in critical areas, and for applying a cleaning/disinfecting solution over a large surface