Imprimer
Chemical resistant gloves, UltraNeo 420
Fournisseur: MAPA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34420347 34420348 34420349 34420020 34420346
112-1081EA 9.5 EUR
112-1081 112-1082 112-1083 112-1084 112-0684
Chemical resistant gloves, UltraNeo 420
Gants
These gloves made from neoprene/natural latex with cotton flock lining provide standard chemical protection.
  • Cotton flock lining for wearing comfort
  • Good grip thanks to embossed finish
  • Extremely supple with freedom of movement
Typical applications include: Automotive/mechanical industry, electric accumulator and battery manufacturing, handling, loading and unloading of chemicals, spreading fertiliser, preparation of industrial glues, industrial cleaning, maintenance.
Certifications: EN 388:2016 (2121X), EN ISO 374-1 Type A (ALMNST), EN ISO 374-5; Cat. III