Imprimer
Cold trap, type KF54V
Fournisseur: KGW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17110 17111 17112 17109 17113
KGWA17110EA 1970 EUR
KGWA17110 KGWA17111 KGWA17112 KGWA17109 KGWA17113
Cold trap, type KF54V
Vacuum Traps
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask