Imprimer
LED spot illuminator, K LED, for stereo microscopes
Fournisseur: ZEISS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4355259010000 4355259020000
ZEIS4355259010000EA 0 EUR
ZEIS4355259010000 ZEIS4355259020000
LED spot illuminator, K LED, for stereo microscopes
Éclairages Éclairage LED
The K LED spot lighting for stereo microscopes stemi 305 and 508.
Spot Illuminator K LED: Single spot for brightfield and darkfield epi-illumination
Thread M24×0,5 for polariser
Height adjustable, inclinable, zoomable
For stands K EDU/LAB/MAT and stand M LED
Double Spot Illuminator K LED: Gooseneck double spot, 2× 160 mm for variable epi-illumination
2× thread M24×0,5 for polariser
Height adjustable
For stands K EDU/LAB/MAT and stand M LED