Imprimer
Dough scraper
Fournisseur: KING DISPOSABLES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
EL2944 EL2945 EL2946
KINGEL2944EA 13.1 EUR
KINGEL2944 KINGEL2945 KINGEL2946
Dough scraper
Raclettes
These dough scrapers feature a white blade made from PE.
  • Spoons, scrapes, and spreads evenly
  • Commercial dishwasher safe
  • Large size perfect for professional kitchens
  • Blade seamlessly attached to handle to reduce dirt and bacteria build-up