Imprimer
Stitch cutter, disposable, 0526
Fournisseur: Swann-Morton
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SWAN0526EA 25 EUR
SWAN0526
Stitch cutter, disposable, 0526
Bistouris et pièces détachées Bistouris
Sterile stitch cutter mounted on a plastic handle allowing extra dexterity and distance from the wound.
  • Stainless steel blade
  • PS handle
Used for pain free removal of modern day suture materials on healing of the wound.