Imprimer
Gas filter connecting units
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1035004 1035008 1035044 1035048 1035024 1035028 1035014 1035018
SGEA1035004EA 547 EUR
SGEA1035004 SGEA1035008 SGEA1035044 SGEA1035048 SGEA1035024 SGEA1035028 SGEA1035014 SGEA1035018
Gas filter connecting units
Porte-filtres
A connecting unit with inlet and outlet connectors for the gas lines.
  • Allow easy installation of gas purifiers into the gas line
  • Unit can be bench or wall mounted
  • Available in 1,2 and 4-port configurations
Gas purifiers are an essential part of your GC analysis as contaminants in gases can significantly impact the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation. Even if carrier gas is of the highest quality, contaminants can be picked up from every part of the gas line. Therefore, a gas purifier is needed to ensure that maximum productivity is achieved.
Informations de commande: Connecting units and filters must be ordered separately.