Imprimer
Examination gloves, nitrile, Sempercare® nitrile skin²
Fournisseur: SEMPERIT TECHN PROD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
826793843 826793845 826793849 826793847
SEMP826793843EA 32.1 EUR
SEMP826793843 SEMP826793845 SEMP826793849 SEMP826793847
Examination gloves, nitrile, Sempercare® nitrile skin²
Gants
These disposable gloves for medicine and industry are made from nitrile butadiene rubber.
  • Non sterile, powder-free
  • Elastic material offers highest wearing comfort and a maximum sense of touch
  • Structured fingertips guarantee a good grip at all times, even in a moist environment
  • Lavender blue colour makes it easy to distinguish from natural latex gloves and allows for easy recognition
  • Do not contain any natural latex proteins: Ideal for people with type I allergy
  • Protect users and patients from the transmission of infections (according to virus impermeability requirements of ASTM F 1671)
The Sempercare® Nitrile Skin² is both labelled as medical device (class 1) and single-use protective glove (PPE category III). Therefore, it can be used for additional applications such as laboratory/pharma, food/catering, professional cleaning/hygiene.
Certifications: EN 455, EN 374, Cat. III (PPE); Class 1 (Medical device); AQL 1,5.