Imprimer
Chromatography tubing, PEEK
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-T-6004-M3 JR-T-60041-M3 JR-T-60042-M3 JR-T-6006 JR-T-6004-M10 JR-T-60042-M10 JR-T-60041-M10
JOURJR-T-6004-M3EA 260 EUR
JOURJR-T-6004-M3 546-1398 546-1399 JOURJR-T-6006 JOURJR-T-6004-M10 JOURJR-T-60042-M10 JOURJR-T-60041-M10
Chromatography tubing, PEEK
Tuyaux Tubulures à chromatographie
PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting, and is not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel.
  • Chemical resistant
The polymer surface will not leach metal ions into the eluent or extract metal-sensitive components from the sample.
Attention: THF and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.