Imprimer
Sample vials, clear, with screw thread, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C40412000A5B2 C40452700B5C2 C40502700C0C2 C40752700D0C2 C40903000F0C2 C40552000B0B2
SCERC40412000A5B2EA 92.2 EUR
SCERC40412000A5B2 SCERC40452700B5C2 391-6132 SCERC40752700D0C2 SCERC40903000F0C2 SCERC40552000B0B2
Sample vials, clear, with screw thread, KIMBLE®
Fioles
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.