Imprimer
SPE columns, Supra-Clean® SCX
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Supra-Clean®
PERCN9306432EA 513 EUR
PERCN9306432 PERCN9306540 PERCN9306538 PERCN9306539 PERCN9306536 PERCN9306537
SPE columns, Supra-Clean® SCX
Colonnes pour chromatographie
Strong Cation Exchange (SCX) Supra-Clean® spherical silica-based SPE columns are recommended for extracting basic compounds when analysing cations, antibiotics, drugs, amino acids, catecholamines, herbicides, nucleic acid bases, nucleosides and surfactants.