Imprimer
Plastic ACME threaded handles
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
STAEH-101WH STAEH-183WH STAH-36WH STAH-48WH STAEH-237BL STAH-24WH STAH-60WH STAEH-072WH STAEH-036WH
MICRSTAEH-101WHEA 279 EUR
MICRSTAEH-101WH 129-1075 MICRSTAH-36WH MICRSTAH-48WH MICRSTAEH-237BL MICRSTAH-24WH MICRSTAH-60WH MICRSTAEH-072WH MICRSTAEH-036WH
Plastic ACME threaded handles
Serpillières
Lightweight, plastic coated fiberglass handles with threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
  • Lightweight, plastic coated fiberglass handles
  • ACME Connect threaded end connection
  • Hand grips available in four color options for colour coding
  • Available fixed length or extendable
  • Fixed lengths range from 300 mm to 1,5 m
  • Extendable maximum lengths range from 900 mm to 3,5 m
  • ACME Connect threaded end attaches directly to plastic threaded mops, tools and adapters
  • SSA-2 adapter connects threaded handles to Quick Disconnect mops, tools and adapters
  • Good compatibility with disinfectants
  • Autoclavable
The Micronova Plastic ACME Threaded Handles attach directly to the plastic (QP series) adapters and tools. Use the SSA-2 ACME Thread Adapter to attach to stainless steel Quick Disconnect (QD series) adapters and tools.