Imprimer
Wipes for general applications, WYPALL* X50, colour coded
Fournisseur: KIMBERLY CLARK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7443 7441 7442 7444
115-2238EA 126 EUR
115-2238 115-1545 115-1546 115-1547
Wipes for general applications, WYPALL* X50, colour coded
Wipes Disposable Wipes and Towels Wipes for General Applications
Due to its HYDROKNIT* material, WYPALL* X50 cloth provides fast absorption, tear resistance and high performance.
  • Best value reusable wiping system for task segregation
  • Good absorption, tear resistant
  • Colour coded and therefore suitable for use in food processing industries
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable