Imprimer
Chemical resistant gloves, Showa 3415
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3415-11 3415-10 3415-08 3415-09
112-5783EA 252 EUR
112-5783 112-5782 112-5780 112-5781
Chemical resistant gloves, Showa 3415
Gants
These seamless knitted gloves with flexible, consistent neoprene coating provide protection against a wide range of chemicals, including acids, caustics, solvents, grease and oil.
  • Fully coated
  • Rough particle grip
  • Gauntlet cuff for protection of the forearm
Recommended industries: Mechanical, automotive, petrochemical.
Certifications: EN 388:2016 (3031X), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5; Cat. III