Imprimer
Test d'ajustement pour protections respiratoire, FT10
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FT10 FT12 FT14
111-0453EA 526 EUR
111-0453 111-0459 111-2539
Test d'ajustement pour protections respiratoire, FT10
Protection respiratoire
Test kits and accessories designed to help your organisation meet OSHA fit testing requirements for respirators. The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapour respirator with a particulate prefilter. FT-10 uses a test solution of sodium saccharin that produces a sweet tasting aerosol.
  • Qualitative fit test kits are suitable for testing wearers of disposable respirators and half face masks fitted with particle filters
  • Kits rely on a ‘taste test’ to assess the level of fit of the respirator to the wearer’s face
Pour les respirateurs 3M : 8812, 9322, 8835, série 4000 série 6000, série 7500.
Certifications: Respirator should be fit tested, as required under the Control of Substances Hazardous to Health Regulations (COSHH). The kit meets the performance criteria for fit testing respirators under the current OSHA Standard for Respirator Protection: 29 CFR 1910.134, Appendix A.
Informations de livraison: Kit includes: hood, collar, 2 nebulisers, sweet tasting sensitivity solution, sweet tasting fit test solution and laminated user instructions.