Imprimer
Lingette M-Zone™ Mittwipes™
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MW-611 MWD-611 MW-AA MW-6 MW-2011 MW-20 MW-11 MW-6IR MWDT-611
112-1293EA 20.9 EUR
112-1293 129-0618 129-0617 129-0616 129-0663 129-0615 MICRMW-11 MICRMW-6IR MICRMWDT-611
Lingette M-Zone™ Mittwipes™
Lingettes Chiffons nettoyants réutilisables Chiffons pour tâches délicates
These cleaning mitts offer the versatility of a high quality wipe with the convenience and security of a glove. Designed for simplified cleaning around hard to reach areas in the cleanroom.
  • Durable cleaning mitts for simplified cleaning
  • Versatility of a high quality wipe with the convenience and security of a glove
  • Dimensions: 300×200 mm
  • For aseptic applications: Available irradiated or can be autoclaved prior to use
  • Available with or without a urethane foam layer
  • Compatible with strong disinfectants
  • Reusable and autoclavable
The Micronova PolyMitt™ Cleaning Mitt Wipes come in one standard size, designed to fit comfortably over a gloved hand, making it easier to clean around carts, shelving, tanks, tubing and other hard to clean areas in the cleanroom.
Available in a range of fabrics for specialised cleaning:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester for residue removal and disinfectant dispersal
MegaTe×™ - 100% non-woven polyamide is absorbent with mild abrasive properties
Microfiber™ - 80% polyester, 20% polyamide is highly absorbent and effective wet or dry