Imprimer
Manchettes de protection résistantes aux produits chimiques, Tychem® 2000 C et Tychem® 6000 F, PS32LA
Fournisseur: DuPont
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TYCPS32SYL00 TYFPS32SGY00
113-0879EA 153 EUR
113-0879 113-0883
Manchettes de protection résistantes aux produits chimiques, Tychem® 2000 C et Tychem® 6000 F, PS32LA
Vêtements à usage unique
Specially designed for use with Tychem® apparel, these sleeve protectors can help offer enhanced protection for body parts that are more exposed to hazardous substances.
  • Wide elastics at cuffs and upper arm
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Anti-static treatment (EN 1149-5) on inside
Tychem® 2000 C garments utilise the strength of Tyvek® and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure). Ideally used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing.
Tychem® 6000 F garments are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents. Ideally used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Certifications: Type PB [3], EN 14126; Cat. III.