Imprimer
Protections de manchons pour salle blanche, BioClean-D™
Fournisseur: Nitritex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S-BDSC-L BDSC-L
113-8035EA 400 EUR
113-8035 113-8034
Protections de manchons pour salle blanche, BioClean-D™
Vêtements à usage unique
These extra long BioClean-D™ disposable sleeve covers are made from CleanTough™ fabric, which is spunbonded non woven PP laminated with a film of PE. This fabric allows comfort and flexibility during use and protection against fine sprays and particles.
  • Lightweight, low linting and hard-wearing material
  • Elasticated arms and cuffs
  • Available as sterile or non sterile version
  • A unique barcode on the outer carton (ITF-14) allows automatic data entry into Stock Control Systems
Suitable for use in Class 10, ISO 4, M2.5 cleanrooms.
Type PB [6]; Cat. III
Certifications: EN ISO 13034 (Type PB[6]), EN 14325, EN 1149-5, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Cdt: Sterile version: One pair per sealed plastic bag, 15 sealed plastic bags (30 pieces) per sealed outer bag, six outer bags (30 pieces) per sealed carton liner, one carton liner per carton (180 pieces / 90 pairs).
Non sterile version: 15 pairs per sealed inner plastic bag, one inner bag per sealed outer bag, six outer bags of 15 pairs per sealed carton liner, one carton liner per carton (90 pairs).