Imprimer
Multi-channel pipettes, manual, variable volume, with LTS™ and RFID tag, Pipet-Lite™ XLS+
Fournisseur: RAININ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17013802 17013803 17013804 17013805 17013806 17013807 MT17013808 17013809 17013810 17013811 MT17014497 17014496
RAIN17013802EA 2080 EUR
RAIN17013802 RAIN17013803 RAIN17013804 RAIN17013805 RAIN17013806 RAIN17013807 JJBEMT17013808 RAIN17013809 RAIN17013810 RAIN17013811 JJBEMT17014497 RAIN17014496
Multi-channel pipettes, manual, variable volume, with LTS™ and RFID tag, Pipet-Lite™ XLS+
Pipettes Pipetteurs
This ergonomic manual pipette with comfortable handle ensure light and smooth operation and significantly reduce the risk of repetitive strain injuries.
  • LTS (LiteTouch System) reduces tip ejection forces by 85% and prevents RSI injuries
  • Effortless pipetting due to light weight, light plunger forces and lowest tip ejection forces
  • Sealing system minimises 'piston stickiness', providing greater control, particularly with applications that require slow, careful pipetting
  • Sophisticated mechanical designs (new push-rod design) ensure high performance, and superior channel-to-channel consistency
  • On-board RFID provides ease of service and calibration tracking
  • Intended for use with LTS™ LiteTouch™ tips