Imprimer
Lysing kits for Fastprep®-24 5G, Lysing Matrix K, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
116920100 116920050
ICNA116920100EA 357 EUR
ICNA116920100 ICNA116920050
Lysing kits for Fastprep®-24 5G, Lysing Matrix K, MP Biomedicals
Homogénéisateurs Homogénéisateurs de paillasse
Each impact-resistant 2 ml tube contains 0,8 mm zirconium silicate beads.
  • Matrix K tubes have brown caps
  • Can be stored at room temperatue
  • DNase and RNase free
Zirconium oxide has very high density and high tensile strength and hardness, and is effective at breaking very tough and hard samples. The small spheres are also effective in breaking fungal spores, cysts, and yeast polysaccharide capsules.