Imprimer
Lysing kits for Fastprep®-24 5G, Lysing Matrix H, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
116917050 116917100 116917500
ICNA116917050EA 225 EUR
ICNA116917050 ICNA116917100 ICNA116917500
Lysing kits for Fastprep®-24 5G, Lysing Matrix H, MP Biomedicals
Homogénéisateurs Homogénéisateurs de paillasse
Each impact-resistant 2 ml tube contains 2 mm glass beads and 2 mm yellow zirconium oxide beads.
  • Used to isolate RNA and proteins from bacterial aggregates, biofilms, and whole insects
  • Matrix H tubes have yellow caps
  • DNase and RNase free
Zirconium oxide has very high density and high tensile strength and hardness, and is effective at breaking very tough and hard samples, including organisms within a tough or dense exterior matrix. This is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing. The 2 mm glass beads are effective at breaking pathogenic yeast polysaccharide capsules and hyphae inside infected tissue.