Imprimer
Lysing kits for Fastprep®-24 5G, Lysing Matrix G, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
116916050 116916100
ICNA116916050EA 225 EUR
ICNA116916050 ICNA116916100
Lysing kits for Fastprep®-24 5G, Lysing Matrix G, MP Biomedicals
Homogénéisateurs Homogénéisateurs de paillasse
Each impact-resistant 2 ml tube contains 1,6 mm silicon carbide particles and 2 mm glass beads.
  • Used in extractions from yeast, fungi and spores, plant and animal tissue and more
  • Matrix G tubes have brown caps
  • Can be stored at room temperature (15 to 30 °C)
  • DNase and RNase free
Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. This is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing. The glass beads are effective at breaking pathogenic yeast polysaccharide capsules and hyphae inside infected tissue.