Imprimer
Taq CORE kits, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11EPTQK101 11EPTQK255C 11EPTQKL255 11EPTQK300 11EPTQK25L 11EPTQK259 11EPTQK251 11EPTQK109 11EPTQK105C 11EPTQK103 11EPTQK253 11EPTQKD251 11EPTQKD253 11EPTQKD259 11EPTQS901K
ICNA11EPTQK101EA 214 EUR
ICNA11EPTQK101 ICNA11EPTQK255C ICNA11EPTQKL255 ICNA11EPTQK300 ICNA11EPTQK25L ICNA11EPTQK259 ICNA11EPTQK251 ICNA11EPTQK109 ICNA11EPTQK105C ICNA11EPTQK103 ICNA11EPTQK253 ICNA11EPTQKD251 ICNA11EPTQKD253 ICNA11EPTQKD259 ICNA11EPTQS901K
Taq CORE kits, MP Biomedicals
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
The Taq CORE kits are designed for the amplification of specific regions of DNA using PCR. The PCR CORE Kits - Direct Loading provide Taq DNA Polymerase, PCR Direct Loading buffers and dNTPs necessary for amplification and subsequent loading of samples on an agarose gel.
  • Contain dNTPs at 10 mM
  • Highly reproducible results
  • Contains ultra-pure enzyme