Imprimer
G418 disulfate solution 50 mg/ml
Marques : Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Geneticin®
30-234-CI 30-234-CR
MDTC30-234-CIEA 566 EUR
MDTC30-234-CI 30-234-CR
G418 disulfate solution 50 mg/ml
G418 disulfate
G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 µg/ml.
This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene. G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.
Attention: For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.
Formule: C₂₀H₄₄N₄O₁₈S₂
Poids moléculaire: 692.72 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00058314
Numéro CAS: 108321-42-2