Imprimer
Semi-automated pipetting system, epMotion® 96
Fournisseur: Eppendorf
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
epMotion®
EPPE5069000.110EA 0 EUR
EPPE5069000.110 EPPE5069000.012 EPPE5069000.217 EPPE5069000.314
Semi-automated pipetting system, epMotion® 96
Automated Liquid Handling Systems
Eppendorf epMotion 96 is an easy to use bench top system for high precision pipetting in 96- and 384-well plates. Its ergonomic design and intuitive handling makes epMotion 96 a great tool for anyone who needs fast and precise liquid handling in a 96-well format.
  • 0,5 to 300 μl with one system without the need to change heads
  • Possibility to use 50 μl or 300 μl tips to maximise accuracy; auto-detection of tip size
  • Compact design to fit under a laminar flow hood; can be easily moved between labs
  • Lifting table supports a variety of SBS-size labware such as micro plates, reservoirs, and Thermoadapters
  • With accessory plate for handling 384-well plates by indexing with the lifting table
  • Tip attachment lever: Minimal force needed to attach tips to the 96 nose-cone array for ergonomic operation
  • Apple® iPod® control unit for convenient system control; accessible by other iOS 7 devices (iPhone®, iPod®) via WiFi
The software is based on Eppendorf Xplorer® electronic pipettes and provided as a free Apple® app. The app provides intelligent, preset applications: aspiration, dilution, sequential dispensing, multi dispensing.
Typical applications include: Replication and reformatting of microplates, cell seeding and media change, reagent and compound addition, 384-wells by 4 times 96-well pipetting, cell-based assays, ELISA handling (plate coating & wash), biochemical assays.
Informations de commande: The system is supplied without iPod® controller. The Apple® iPod controller must be ordered separately.