Imprimer
Stoppers, rubber, long series with slight taper
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
201024 201058 201082 201014
217-0513EA 22.7 EUR
217-0513 217-9479 217-9486 217-9476
Stoppers, rubber, long series with slight taper
Stoppers Rubber Stoppers
Conical natural grey-blue rubber stoppers without holes.
  • Resistant to common laboratory chemicals
  • Highly elastic, for a perfect seal and exceptional bonding
  • Resistant to temperatures from −5 to +70 °C (+120 °C in humid atmosphere)
Resistant to temperatures from -5 to +70 °C (+120 °C in humid atmosphere)