Imprimer
pH electrode, pHenomenal®
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
620-1659EA 186 EUR
620-1659 662-1162 662-1163 662-1164 662-1158 662-1159 662-1161 662-1156 662-1157 662-1247
pH electrode, pHenomenal®
Electrodes pH Electrodes
pH electrodes with BNC connector and cable.
These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.
* Integrated temperature probe.
Informations de livraison: Includes pH-electrode only. 620-1659 includes pHenomenal® TEMP21 T-sensor (NTC 30 kOhm) only.