Imprimer
Gants en cotte de maille résistants aux coupures, Chainex 2000
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2500000R0302 2500001R0302 2500002R0302 2500003R0302 2500004R0302 2500005R0302
112-1243EA 156 EUR
112-1243 112-1244 112-1249 112-1255 112-1256 112-1257
Gants en cotte de maille résistants aux coupures, Chainex 2000
Gants
The Chainex 2000 is a full stainless steel glove with short cuff and attached textile strap. Chainex 2000 is a fully reversible product range, and is equipped with heavy duty adjustable straps and press stud fastening. Offers excellent cut and puncture protection. The high quality of the chain mesh reduces the numbers of repairs, and extends the life time of the glove.
  • Smooth textile straps adapt to all wrist profiles
  • Fully reversible gloves can be used on left or right hand
  • Wrist fastens with press stud and adjustable buckle
Stainless chainmail thickness: 0,5 mm.
External diameter of the rings: 4,0 mm.
Weight: 129 to 219 g (depending on size).
Certifications: EN 1082, Cat. II.