Imprimer
Avantor® Apollo C18, HPLC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HICH36505EA 673 EUR
HICH36505 HICH36515 HICH36511 HICH36521 HICH970205 HICH36543 HICH36537 96454/N
Avantor® Apollo C18, HPLC Columns
Colonnes pour chromatographie
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
  • Easy scale-up from analytical to preparative
  • Extended pH stability is about 1,5 to 10,5
  • USP L1
These columns are ideal for routine analysis and for educational laboratories.