Imprimer
Dehydrated culture media, Circlegrow®, Molecular Biology Certified™, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
113000132 113000131 113000142 113000112 113000121 113000122 113000136 113000152
ICNA113000132EA 243 EUR
ICNA113000132 ICNA113000131 ICNA113000142 ICNA113000112 ICNA113000121 ICNA113000122 ICNA113000136 ICNA113000152
Dehydrated culture media, Circlegrow®, Molecular Biology Certified™, MP Biomedicals
Milieux pour microbiologie Milieux pour microbiologie déshydratés
Circlegrow® is designed for obtaining rapid growth of E. coli and high yields of plasmid DNA. It extends plasmid production past log phase.
  • Available as capsules, powders or in pouches
  • Molecular biology certified
At the end of log phase, cells growing in Circlegrow® begin to utilize a proprietary second carbon source that becomes available on a delayed basis. Cell growth and plasmid replication continue long after cells in conventional media have reached stationary phase or expired.
Bacteria commonly grow in Circlegrow® to a density triple that achieved in other media. The slow post-log phase growth of bacteria in it eliminates the need for constant OD monitoring. These cultures left overnight or over the weekend will have a much lower percentage of dead cells, and a higher percentage of plasmid DNA, than cultures prepared with any other media. The result is a higher yield of plasmid DNA in less volume with less attention.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Certifications: Molecular Biology Certified™.