Imprimer
Racks for test tubes
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2497902 1488600
HACH2497902EA 43.1 EUR
HACH2497902 HACH1488600
Racks for test tubes
Portoirs Tube Racks
These test tube racks features a special blend of polyethylene which sinks in water, assuring non-floating racks when immersed.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance