Imprimer
Pipettes monocanal, mécaniques, à volume fixe, PIPETMAN® L
Fournisseur: GILSON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FA10018 FA10017 FA10019 FA10023 FA10024 FA10025 FA10026 FA10027 FA10028 FA10029 FA10030 FA10031 FA10020 FA10021 FA10022
GILSFA10018EA 195 EUR
GILSFA10018 GILSFA10017 GILSFA10019 GILSFA10023 GILSFA10024 GILSFA10025 GILSFA10026 GILSFA10027 GILSFA10028 GILSFA10029 GILSFA10030 GILSFA10031 GILSFA10020 GILSFA10021 GILSFA10022
Pipettes monocanal, mécaniques, à volume fixe, PIPETMAN® L
Pipettes Pipetteurs
The PIPETMAN® L Fixed pipettes offer the same comfortable, lightweight design as the variable volume models with a fixed volume that reduces the risk of any volume error during pipetting. This makes them an ideal choice for clinical diagnostics, quality control, and any routine testing applications even for non-trained lab technicians.
  • Comfortable pipetting
  • GLP compliance
Based on the ergonomic design of the original PIPETMAN L, the fixed volume model offers an ideal weight and handle shape. The more natural fit of the pipette lets you relax your grip, making it a more ergonomic option for your hand while pipetting. It also provides an adjustable tip ejector so it's easy to switch for either right- or left-handed users, whichever is most comfortable. The graduated cover can be readjusted by users for dense or viscous fluids (once reset, the new volume cannot be accidentally changed).
PIPETMAN L variable volume models are all fully autoclavable without disassembly for maximum convenience and protection from cross-contamination. PIPETMAN L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.