Imprimer
MXX low pressure selection valves, Rheodyne®, MX Series II™
Fournisseur: IDEX RHEODYNE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MXX777-605 MXX778-605
554-2671EA 4250 EUR
554-2671 554-3289
MXX low pressure selection valves, Rheodyne®, MX Series II™
Vannes Vannes de sélection
Combine MX Series II™ modular valves with an instrument currently in use to support complex fluid switching and sample injection needs.
  • Can withstand up to 125 psi (9 bar)
  • All models accept 1/16" or 1/8" tubing (proper ferrules required)
  • Multiple control options
  • Stand alone capability
  • Included software