Imprimer
Plastic hydrometers, Durac™, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B618000100 B618000000
BELAB618000100EA 413 EUR
BELAB618000100 BELAB618000000
Plastic hydrometers, Durac™, SP Bel-Art
Hydromètres
Made from optical quality polycarbonate, these plain form hydrometers are designed for applications where rough handling or contamination could be an issue.
  • Durable and shatterproof, lead free ballast
  • Easy to read black scale on yellow back to avoid eye fatigue
  • Individually serialised − Temperature standardisation of 15 °C
Suitable for applications in the food or beverage industries.
Certifications: Traceable to National Institute of Standards and Technology (NIST).
Informations de livraison: Includes a multi-language statement of accuracy.
Attention: Not recommended for use with petroleum-based liquids or other corrosives, these should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.