J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Marques : AvantorTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Silica-based media used for the purification of proteins based on differential salt induced hydrophobicity. Mixed mode provides unique selectivity with its hydrophobic interaction and ion exchange function.