Imprimer
Cation exchange chromatography columns, TSK-GEL® SCX
Fournisseur: TOSOH Bioscience
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0007156 0007158
TOYO0007156EA 1890 EUR
TOYO0007156 TOYO0007158
Cation exchange chromatography columns, TSK-GEL® SCX
Colonnes pour chromatographie
TSK-Gel® SCX are polymer based stainless steel columns packed with porous polystyrene divinylbenzene polymer beads. Designed for the separation and analysis of organic acids, saccharides and alcohols.